2-Chloro-4-methyl-5-nitropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-5-nitro-4-picoline

361030010 361030050
TS361030010EA 35 USD
TS361030010 TS361030050
2-Chloro-4-methyl-5-nitropyridine 98%
2-Chloro-4-methyl-5-nitropyridine
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Boiling Pt: 91 °C (5 mmHg)
Melting Pt: 36…40 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00010688
CAS Number: 23056-33-9
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Cream to light brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 36 to 40 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR