5-Bromo-2-chloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

360670010 360670050 360670250
TS360670010EA 19 USD
TS360670010 TS360670050 TS360670250
5-Bromo-2-chloropyridine 98%
5-Bromo-2-chloropyridine
Formula: C₅H₃BrClN
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD01318951
CAS Number: 53939-30-3

Order Now

Specification Test Results

Appearance White to yellow to brown Crystalline powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 67 to 72 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR