Silica gel, functionalized, Amino-3 ∼1.4 mmol/g, Diameter 40-63 micron

Supplier: Thermo Scientific Chemicals

360180250 360181000 360180100
TS360180250EA 124 USD
TS360180250 TS360181000 TS360180100
Silica gel, functionalized, Amino-3 ∼1.4 mmol/g, Diameter 40-63 micron
Silica gel, functionalized, Amino-3
Formula: C₉H₂₃NO₅Si₂
Storage Temperature: Ambient
MDL Number: MFCD03427643
CAS Number: 71888-97-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size 40 to 63 µm
Loading ≥1.2 mmol/g (molecular)
Nitrogen (N) ≥1.68 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR