3-(Trifluoromethoxy)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-(Trifluoromethoxy)phenyl)metaboric acid, (3-(Trifluoromethoxy)phenyl)boronic acid, 3-(Trifluoromethoxy)benzeneboronic acid, Dihydroxy(3-(trifluoromethoxy)phenyl)borane

359720010 359720050
TS359720010EA 19 USD
TS359720010 TS359720050
3-(Trifluoromethoxy)phenylboronic acid 98%
3-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
Melting Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD01320697
CAS Number: 179113-90-7

Order Now

Specification Test Results

Appearance White to light beige Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR