4-Ethylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

359320010 359320050
TS359320010EA 15 USD
TS359320010 TS359320050
4-Ethylphenylboronic acid 97%
4-Ethylphenylboronic acid
Formula: C₈H₁₁BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00859377
CAS Number: 63139-21-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR