Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

355180010 355180050
TS355180010EA 34 USD
TS355180010 TS355180050
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate 98%
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00051652
CAS Number: 31037-02-2

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 98 to 102 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR