(3-Fluorophenyl)acetone 98%

Supplier: Thermo Scientific Chemicals

350960010 350960050
TS350960010EA 27 USD
TS350960010 TS350960050
(3-Fluorophenyl)acetone 98%
(3-Fluorophenyl)acetone
Formula: C₉H₉FO
Storage Temperature: Ambient
MDL Number: MFCD00061144
CAS Number: 1737-19-5

Order Now

Specification Test Results

Appearance Clear yellow-green Liquid
Infrared spectrum Conforms
Refractive index 1.4955 to 1.4975 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR