Ethyl-2-morpholinoacetate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl 2-morpholinoacetate, Ethyl morpholinoacetate

350910050 350910250
TS350910050EA 16 USD
TS350910050 TS350910250
Ethyl-2-morpholinoacetate 98%
Ethyl-2-morpholinoacetate
Formula: C₈H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00023375
CAS Number: 3235-82-3
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4535 to 1.4555 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR