2-Amino-5-bromobenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

350250010 350250050
TS350250010EA 37 USD
TS350250010 TS350250050
2-Amino-5-bromobenzonitrile 97%
2-Amino-5-bromobenzonitrile
Formula: C₇H₅BrN₂
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00158946
CAS Number: 39263-32-6
EINECS: 254-387-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 93 to 98 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR