4-Chloro-6-methoxypyrimidin-2-amine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-4-chloro-6-methoxypyrimidine

347090050 347090250
TS347090050EA 29 USD
TS347090050 TS347090250
4-Chloro-6-methoxypyrimidin-2-amine 95%
4-Chloro-6-methoxypyrimidin-2-amine
Formula: C₅H₆ClN₃O
MW: 159.57 g/mol
Melting Pt: 169…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00233535
CAS Number: 5734-64-5

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 169 to 176 °C
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR