tert-Butyl (3R)-3-amino-5-methylhexanoate 97%

Supplier: Thermo Scientific Chemicals
Danger

337925000 337921000
TS337925000EA 231 USD
TS337925000 TS337921000
tert-Butyl (3R)-3-amino-5-methylhexanoate 97%
tert-Butyl (3R)-3-amino-5-methylhexanoate
Formula: C₁₁H₂₃NO₂
Storage Temperature: Freezer
MDL Number: MFCD01076257
CAS Number: 166023-29-6
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4330 (20 °C, 589 nm)
GC ≥96.0 %
Specific optical rotation -11° to -15° (20 °C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR