(2-Pyrimidylthio)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

334490050 334490250
TS334490050EA 30 USD
TS334490050 TS334490250
(2-Pyrimidylthio)acetic acid 98%
(2-Pyrimidylthio)acetic acid
Formula: C₆H₆N₂O₂S
Melting Pt: 200…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00010287
CAS Number: 88768-45-0

Order Now

Specification Test Results

Appearance Beige to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR