Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
325710010
325710050
TS325710010EA
25
USD
TS325710010
TS325710050
Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate 95%
Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate
Formula:
C₁₂H₁₂O₅ Melting Pt: 81…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00800637 CAS Number: 22027-52-7 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Off-white to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 81 to 86 °C |
Assay | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...