2-Hydroxy-5-nitro-4-picoline 98%

Supplier: Thermo Scientific Chemicals
Warning

319570010 319570050
TS319570010EA 17 USD
TS319570010 TS319570050
2-Hydroxy-5-nitro-4-picoline 98%
2-Hydroxy-5-nitro-4-picoline
Formula: C₆H₆N₂O₃
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00010690
CAS Number: 21901-41-7

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 186 to 190 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR