Indium 99.99% (trace metals basis), foil, Thickness: 0.127 mm

Supplier: Thermo Scientific Chemicals

318110023 318110092
TS318110023EA 154 USD
TS318110023 TS318110092
Indium 99.99% (trace metals basis), foil, Thickness: 0.127 mm
Indium
Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now

Specification Test Results

Appearance Grey Foil
Total trace metal impurities ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR