Barium 99+% in mineral oil, rods, Diameter ∼ 2 cm

Supplier: Thermo Scientific Chemicals
Danger

317861000 317865000
TS317861000EA 234 USD
TS317861000 TS317865000
Barium 99+% in mineral oil, rods, Diameter ∼ 2 cm
Barium
Formula: Ba
Boiling Pt: 1640 °C
Melting Pt: 725 °C
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Grey Rods
Assay ≥99.0 % Ba (incl. Sr)
Strontium (Sr) ≤0.8 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR