Tin 99.8%, powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

317801000 317805000
TS317801000EA 43 USD
TS317801000 TS317805000
Tin 99.8%, powder -325 mesh
Tin
Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.3 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now

Specification Test Results

Appearance Grey Powder
Total trace metal impurities ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR