3,5-Dichloro-2,4,6-trifluoropyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,5-Dichlorotrifluoropyridine

311400050 311400250
TS311400050EA 34 USD
TS311400050 TS311400250
3,5-Dichloro-2,4,6-trifluoropyridine 98%
3,5-Dichloro-2,4,6-trifluoropyridine
Formula: C₅Cl₂F₃N
MW: 201.96 g/mol
Boiling Pt: 159…160 °C (760 mmHg)
Melting Pt: 24…26 °C
Density: 1.622 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006226
CAS Number: 1737-93-5
EINECS: 217-088-3
UN: 3382
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR