Allylpentafluorobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

310850050 310850250
TS310850050EA 41 USD
TS310850050 TS310850250
Allylpentafluorobenzene 97%
Allylpentafluorobenzene
Formula: (C₆F₅)CH₂CHCH₂
Boiling Pt: 148…149 °C
Melting Pt: –64 °C
Density: 1.358 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00000301
CAS Number: 1736-60-3
EINECS: 217-083-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4290 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR