3-(Trifluoromethyl)benzenesulfonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

306280010 306280050
TS306280010EA 18 USD
TS306280010 TS306280050
3-(Trifluoromethyl)benzenesulfonyl chloride 95%
3-(Trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₂S
Boiling Pt: 77…79 °C (.8 mmHg)
Density: 1.52 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00014724
CAS Number: 777-44-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4847 to 1.4867 (20 °C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR