Samarium 99.9% (trace metals basis) 20-40 mesh

Supplier: Thermo Scientific Chemicals
Danger

294780100 294780500
TS294780100EA 208 USD
TS294780100 TS294780500
Samarium 99.9% (trace metals basis) 20-40 mesh
Samarium
Formula: Sm
MW: 150.36 g/mol
Boiling Pt: 1778 °C (760 mmHg)
Melting Pt: 1072 °C
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
UN: 3132
ADR: 4.3,II
Merck Index: 12,08498

Order Now

Specification Test Results

Appearance Silver grey Powder or filings
Particle size 20 to 40 mesh
Total trace metal impurities ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR