4-Chloro-2-methoxyphenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chlor-2-hydroxyanisole, 4-Chloroguaiacol

294110050 294110100
TS294110050EA 56 USD
TS294110050 TS294110100
4-Chloro-2-methoxyphenol 99%
4-Chloro-2-methoxyphenol
Formula: C₇H₇ClO₂
MW: 158.58 g/mol
Boiling Pt: 241 °C (760 mmHg)
Melting Pt: 16…17 °C
Density: 1.3 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00070774
CAS Number: 16766-30-6
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear light yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5575 to 1.5595 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR