4-Bromo-3-methylanisole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene
294030050
294030250
TS294030050EA
34
USD
TS294030050
TS294030250
4-Bromo-3-methylanisole 97%
4-Bromo-3-methylanisole
Formula:
C₈H₉BrO MW: 201.06 g/mol Boiling Pt: 111…112 °C (15 mmHg) Density: 0.91 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060687 CAS Number: 27060-75-9 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5600 to 1.5620 (20 °C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...