5-Nitro-o-toluic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

276570050 276570250
TS276570050EA 74 USD
TS276570050 TS276570250
5-Nitro-o-toluic acid 99+%
5-Nitro-o-toluic acid
Formula: C₈H₇NO₄
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00007371
CAS Number: 1975-52-6
EINECS: 217-829-0

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 176 to 180 °C
Titration with TBAH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR