Zinc-copper couple

Supplier: Thermo Scientific Chemicals
Warning

271441000 271440250
TS271441000EA 260 USD
TS271441000 TS271440250
Zinc-copper couple
Zinc-copper couple
Formula: CuZn
Melting Pt: 500 °C
Storage Temperature: Ambient
MDL Number: MFCD00084851
CAS Number: 53801-63-1
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Grey to black Powder
Copper (Cu) ≤2 %
Zinc (Zn) ≥98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR