N-Methylhippuric acid 99%

Supplier: Thermo Scientific Chemicals
Warning

259760050 259760250
TS259760050EA 93 USD
TS259760050 TS259760250
N-Methylhippuric acid 99%
N-Methylhippuric acid
Formula: C₁₀H₁₁NO₃
Melting Pt: 101…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00144940
CAS Number: 2568-34-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 101 to 105 °C
Titration with TBAH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR