2-Methyl-5-isopropylaniline (synthetic) ≥94% (by GC)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Isopropyl-2-methylaniline, 2-Amino-p-cymene, 5-Isopropyl-o-toluidine

248790010 248790050 248790250
TS248790010EA 116 USD
TS248790010 TS248790050 TS248790250
2-Methyl-5-isopropylaniline (synthetic) ≥94% (by GC)
2-Methyl-5-isopropylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Density: 0.95 g/cm³
MDL Number: MFCD00044503
CAS Number: 2051-53-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown liquid
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5450 (20°C, 589 nm)
Water ≤2.5 %
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR