2-Chloro-3-(trifluoromethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

241790250 241790050
TS241790250EA 41 USD
TS241790250 TS241790050
2-Chloro-3-(trifluoromethyl)pyridine 97%
2-Chloro-3-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
Boiling Pt: 166…168 °C (760 mmHg)
Melting Pt: 36…40 °C
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00042223
CAS Number: 65753-47-1
EINECS: 424-520-6
UN: 2923
ADR: 8 II

Order Now

Specification Test Results

Appearance Semitransparent Crystals
Infrared spectrum Conforms
Melting point 37 to 41 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR