Silica gel, Pore size: 90 Å 0.035-0.070 mm for chromatography

Supplier: Thermo Scientific Chemicals

241650010 241652500
TS241650010EA 138 USD
TS241650010 TS241652500
Silica gel, Pore size: 90 Å 0.035-0.070 mm for chromatography
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16

Order Now

Specification Test Results

Appearance White Powder
Particle size ≤10 % oversized particles
Bulk density 385 to 440 g/l
pH 6.5 to 7.5 (5 % w/w suspension)
Residual water 2 to 4 % (160°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR