Polyvinylpyrrolidone cross-linked

Supplier: Thermo Scientific Chemicals

Synonyms: Polyvinylpolypyrrolidone, N-Vinyl-2-Pyrrolidone

227480010 227480250 227481000 227485000
TS227480010EA 348 USD
TS227480010 TS227480250 TS227481000 TS227485000
Polyvinylpyrrolidone cross-linked
Polyvinylpyrrolidone cross-linked
Formula: C₆H₉NO
MW: 111.144 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149016
CAS Number: 25249-54-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤5 % (K.F.)
Loss on drying ≤5 %
Heavy metals ≤10 ppm
Sulfated ash ≤0.1 %
pH 5 to 8 (1 % in water)
Arsenic (As) ≤2 ppm
Nitrogen (N) 11 to 12.8 % (as is)
Impurity ≤5 ppm free vinyl pyrrolidone
Soluble matter ≤1 % (in water)
Peroxides ≤400 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR