5-Nitrouracil 99+%

Supplier: Thermo Scientific Chemicals
Warning

226580050 226581000
TS226580050EA 15 USD
TS226580050 TS226581000
5-Nitrouracil 99+%
5-Nitrouracil
Formula: C₄H₃N₃O₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006021
CAS Number: 611-08-5
EINECS: 210-250-4

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder or needles
Infrared spectrum Conforms
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR