3,5-Diiodo-L-tyrosine dihydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-Tyr(3,5-DiI)-OH·2H₂O, (S)-2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate

225610050 225610250
TS225610050EA 32 USD
TS225610050 TS225610250
3,5-Diiodo-L-tyrosine dihydrate 99%
3,5-Diiodo-L-tyrosine dihydrate
Formula: C₉H₉I₂NO₃·2H₂O
MW: 469.01 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00016542
CAS Number: 18835-59-1

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Water 6 to 9 % (K.F.)
Specific optical rotation -17° to -20.7° (20 °C, 589 nm) (c=2.5, 0.1N NaOH/H2O) (on dry substance)
Titration with HClO4 ≥98.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR