Talc Ø -200 mesh (<75 µm)

Supplier: Thermo Scientific Chemicals

222730025 222730100 222735000
TS222730025EA 232 USD
TS222730025 TS222730100 TS222735000
Talc Ø -200 mesh (<75 µm)
Talc
Formula: Mg₃O₁₁Si₄·1H₂O
Melting Pt: 800 °C
Storage Temperature: Ambient
MDL Number: MFCD00792903
CAS Number: 14807-96-6
EINECS: 238-877-9
Merck Index: 13,09127

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Particle size ≤75 µm
Loss on ignition ≤7.0 % (1 g, 1100°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR