3-Amino-4-chlorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

219300250 219301000
TS219300250EA 27 USD
TS219300250 TS219301000
3-Amino-4-chlorobenzoic acid 98%
3-Amino-4-chlorobenzoic acid
Formula: C₇H₆ClNO₂
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 214…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00007671
CAS Number: 2840-28-0
EINECS: 220-635-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Off-white to beige to light brown Powder
Infrared spectrum Conforms
Melting point 213 to 216 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR