Cesium 99.95+% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
219190010
219190050
TS219190010EA
166
USD
TS219190010
TS219190050
Cesium 99.95+% (trace metals basis)
Cesium
Formula:
Cs MW: 132.91 Boiling Pt: 705 Melting Pt: 28.5 Density: 1.873 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00134037 CAS Number: 7440-46-2 EINECS: 231-155-4 UN: 1407 ADR: 4.3 I Merck Index: 12,02051 |
Specification Test Results
Appearance | Golden Metal |
Total trace metal impurities | ≤500 ppm |
Cesium (Cs) | ≥99.95 % (metals basis) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...