Bole white (kaolin), pure

Supplier: Thermo Scientific Chemicals

Synonyms: China Clay, Hydrated aluminum silicate, Clay

211740010 211740025 211740100
TS211740010EA 113 USD
TS211740010 TS211740025 TS211740100
Bole white (kaolin), pure
Bole white (kaolin)
Formula: Al₂SiO₇·2H₂O
MW: 258.16 g/mol
Melting Pt: 1760
Storage Temperature: Ambient
MDL Number: MFCD00062311
CAS Number: 1332-58-7
EINECS: 310-194-1
REACH: exempt
Merck Index: 13,05300

Order Now

Specification Test Results

Appearance Light beige Powder
Loss on drying ≤1.5 %
Particle size 1.0 to 1.8 µm (D50)
Loss on ignition 10.8 % to 11.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR