3-Amino-o-cresol 95%

Supplier: Thermo Scientific Chemicals
Warning

209042500 209040010
TS209042500EA 19 USD
TS209042500 TS209040010
3-Amino-o-cresol 95%
3-Amino-o-cresol
Formula: C₇H₉NO
Melting Pt: 127…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00007787
CAS Number: 53222-92-7
EINECS: 258-438-5

Order Now

Specification Test Results

Appearance Brown to black Crystalline powder
Infrared spectrum Conforms
Melting point 127 to 133 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR