(4-Hydroxy-3-nitrophenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

204890010 204890050
TS204890010EA 40 USD
TS204890010 TS204890050
(4-Hydroxy-3-nitrophenyl)acetic acid 99%
(4-Hydroxy-3-nitrophenyl)acetic acid
Formula: (C₆H₃(OH)(NO₂))CH₂COOH
Melting Pt: 144…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00007122
CAS Number: 10463-20-4
EINECS: 233-948-0

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 144 to 147 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR