Calcium 99%, pieces
Supplier: Thermo Scientific Chemicals
|
Danger
|
201180050
201185000
TS201180050EA
47
USD
TS201180050
TS201185000
Calcium 99%, pieces
Calcium
Formula:
Ca Boiling Pt: 1484 °C (1013 hPa) Melting Pt: 845 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085314 CAS Number: 7440-70-2 EINECS: 231-179-5 UN: 1401 ADR: 4.3,II |
Specification Test Results
Appearance | Grey Metallic pieces |
Assay | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...