Palladium hydroxide (20% Pd) on carbon (Pearlman's catalyst) wetted with ca. 60% water, powder, unreduced

Supplier: Thermo Scientific Chemicals

199620500 199620100
TS199620500EA 2866 USD
TS199620500 TS199620100
Palladium hydroxide (20% Pd) on carbon (Pearlman's catalyst) wetted with ca. 60% water, powder, unreduced
Palladium hydroxide
Formula: H₂O₂Pd
Storage Temperature: Ambient
MDL Number: MFCD00064599
CAS Number: 12135-22-7

Order Now

Specification Test Results

Appearance Black Powder
Water 45.0 to 55.0 %
Palladium (Pd) 18.0 to 22.0 % On dry basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR