Iridium 99.9% (trace metals basis), powder

Supplier: Thermo Scientific Chemicals
Danger

198090100 198095000
TS198090100EA 6036 USD
TS198090100 TS198095000
Iridium 99.9% (trace metals basis), powder
Iridium
Formula: Ir
MW: 192.22
Boiling Pt: 5300
Melting Pt: 2450
Storage Temperature: Ambient
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1 II
Merck Index: 12,05098

Order Now

Specification Test Results

Appearance Grey Powder
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR