Gallium 99.9999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

193070010 193070050 193070250
TS193070010EA 38 USD
TS193070010 TS193070050 TS193070250
Gallium 99.9999% (trace metals basis)
Gallium
Formula: Ga
MW: 69.72 g/mol
Boiling Pt: 2403
Melting Pt: 29
Density: 5.9 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8 III
Merck Index: 12,04364

Order Now

Specification Test Results

Appearance Silver white to silver grey Metallic solid or liquid
Total trace metal impurities ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR