α-Chloro-2-methyl-3-nitrotoluene 97%

Supplier: Thermo Scientific Chemicals
Danger

187400010 187400050
TS187400010EA 16 USD
TS187400010 TS187400050
α-Chloro-2-methyl-3-nitrotoluene 97%
α-Chloro-2-methyl-3-nitrotoluene
Formula: C₈H₈ClNO₂
Melting Pt: 42…47 °C
Flash Pt: >109 °C
Storage Temperature: Ambient
MDL Number: MFCD00007165
CAS Number: 60468-54-4
EINECS: 262-249-3
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow-brown Chunks
Infrared spectrum Conforms
Melting point 42 to 47 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR