Poly(acrylic acid), M.W. ∼ 2,000 63% (w/w) in water

Supplier: Thermo Scientific Chemicals

184990050 184992500 184995000
TS184990050EA 26 USD
TS184990050 TS184992500 TS184995000
Poly(acrylic acid), M.W. ∼ 2,000 63% (w/w) in water
Poly(acrylic acid)
Formula: C₃H₄O₂
Boiling Pt: 100 °C (760mmHg)
Density: 1.23 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00084394
CAS Number: 9003-01-4

Order Now

Specification Test Results

Appearance Clear to hazy liquid
Infrared spectrum Conforms
Assay 60.0 to 65.0 % Solids
Molecular weight 2000 (approx)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR