3,5-Dichlorobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

182770250 182770050
TS182770250EA 227 USD
TS182770250 TS182770050
3,5-Dichlorobenzyl alcohol 98%
3,5-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 76…80 °C
MDL Number: MFCD00004634
CAS Number: 60211-57-6
EINECS: 262-109-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥97.5 %
HPLC ≥97.5 %
Specific optical rotation -76° to -82° (20 °C, 589 nm) (2 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR