(S)-(+)-2-Amino-3-methyl-1-butanol 96%

Supplier: Thermo Scientific Chemicals
Warning

181590010 181590050 181590250
TS181590010EA 39 USD
TS181590010 TS181590050 TS181590250
(S)-(+)-2-Amino-3-methyl-1-butanol 96%
(S)-(+)-2-Amino-3-methyl-1-butanol
Formula: C₅H₁₃NO
Boiling Pt: 81 °C (8 mmHg)
Melting Pt: 30…32 °C
Density: 0.92 g/cm³
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064296
CAS Number: 2026-48-4
EINECS: 217-975-5

Order Now

Specification Test Results

Appearance Clear colorless to white to light yellow Liquid after melting or solid
Infrared spectrum Conforms
Refractive index 1.4535 to 1.4565 (20°C, 589 nm)
GC ≥95 %
Specific optical rotation +15° to +19° (20°C, 589 nm) (c=10, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR