PTFE

Supplier: Thermo Scientific Chemicals

178930050 178931000 178932500
TS178930050EA 39 USD
TS178930050 TS178931000 TS178932500
PTFE
PTFE
Formula: (CF₂CF₂)n
Boiling Pt: >400 °C (760 mmHg)
Melting Pt: 327 °C
Density: 2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00084455
CAS Number: 9002-84-0

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥97.5 %
Residue after ign. (sulfated) ≤0.25 %
Specific optical rotation -33° to -37° (20 °C, 589 nm) (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR