Poly(styrene), M.W. ∼ 260,000

Supplier: Thermo Scientific Chemicals

178890010 178890250 178891000 178895000
TS178890010EA 236 USD
TS178890010 TS178890250 TS178891000 TS178895000
Poly(styrene), M.W. ∼ 260,000
Poly(styrene)
Formula: C₈H₈
MW: 125000 - 250000 g/mol
Melting Pt: 93 °C
Density: 1.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00084450
CAS Number: 9003-53-6

Order Now

Specification Test Results

Appearance Clear Pellets
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR