Poly(methyl methacrylate), M.W. ∼35000, beads

Supplier: Thermo Scientific Chemicals

178760010 178760250 178765000
TS178760010EA 269 USD
TS178760010 TS178760250 TS178765000
Poly(methyl methacrylate), M.W. ∼35000, beads
Poly(methyl methacrylate)
Formula: C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
Melting Pt: 105 °C
Flash Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00134349
CAS Number: 9011-14-7

Order Now

Specification Test Results

Appearance White Beads
Infrared spectrum Conforms
Loss on drying ≤5 %
Particle size 50 to 150 microns (approximately)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR