Poly(acrylic acid), M.W. 1080000 (MN 135000) secondary standard

Supplier: Thermo Scientific Chemicals

178060010 178060100
TS178060010EA 34 USD
TS178060010 TS178060100
Poly(acrylic acid), M.W. 1080000 (MN 135000) secondary standard
Poly(acrylic acid)
Formula: C₃H₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00084394
CAS Number: 9003-01-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤10 %
Solubility (2.5 % in pyridine) clear colorless to light yellow
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR